siRNA Details
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 9 | 1135 | ||
NM_001303257.2 | NP_001290186.1 | 9 | 1135 | |||
NM_014941.3 | NP_055756.1 | 10 | 2001 | |||
XM_011530004.1 | XP_011528306.1 | 9 | 1360 | |||
XM_017028667.1 | XP_016884156.1 | 9 | 1360 | |||
XM_017028668.1 | XP_016884157.1 | 10 | 1455 | |||
GenBank mRNA | AB020659.1 | - | 10 | 1915 | ||
BC019257.1 | - | 8 | 662 | 3503651 | ||
BC136782.1 | - | 10 | 860 | 9020772 | ||
CR456469.1 | - | 8 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
