siRNA Details
Gene Symbol: HMGXB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003681.2 | NP_001003681.1 | 11 | 3902 | ||
NR_027780.1 | 12 | 4017 | ||||
XM_006724100.3 | XP_006724163.1 | 10 | 3981 | |||
XM_006724101.3 | XP_006724164.1 | 9 | 3858 | |||
XM_006724102.1 | XP_006724165.1 | 11 | 3885 | |||
GenBank mRNA | AJ010069.1 | - | 6 | 2513 | ||
AL079310.1 | - | 12 | 3966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
