siRNA Details
Gene Symbol: SYNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004711.4 | NP_004702.2 | 2 | 198 | ||
NM_145731.3 | NP_663783.1 | 2 | 198 | |||
NM_145738.2 | NP_663791.1 | 2 | 164 | |||
GenBank mRNA | AJ002303.1 | - | 2 | 164 | ||
AJ002304.1 | - | 2 | 136 | |||
AJ002305.1 | - | 2 | 136 | |||
AK289507.1 | - | 2 | 145 | |||
AK295863.1 | - | 2 | 156 | |||
BC000731.2 | - | 2 | 162 | 3508457 | ||
BI667990.1 | - | 2 | 222 | |||
BT007135.1 | - | 2 | 136 | |||
CR456590.1 | - | 2 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
