siRNA Details
Gene Symbol: L3MBTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031488.4 | NP_113676.2 | 14, 15 | 1884 | ||
XM_011530420.2 | XP_011528722.1 | 13, 14 | 1867 | |||
XM_011530421.2 | XP_011528723.1 | 14, 15 | 1953 | |||
XM_017028976.1 | XP_016884465.1 | 12, 13 | 1661 | |||
XM_017028977.1 | XP_016884466.1 | 13, 14 | 2180 | |||
XM_017028978.1 | XP_016884467.1 | 14, 15 | 2135 | |||
XM_017028979.1 | XP_016884468.1 | 14, 15 | 2080 | |||
XM_017028980.1 | XP_016884469.1 | 13, 14 | 1767 | |||
GenBank mRNA | AJ305226.1 | - | 14, 15 | 1869 | ||
AJ305227.1 | - | 15, 16 | 1974 | |||
AK056443.1 | - | 14, 15 | 1881 | |||
AK074091.1 | - | 13 | 4568 | |||
AL136564.1 | - | 14 | 2364 | |||
BC017191.2 | - | 14, 15 | 1873 | 3138444 | ||
BU184979.1 | - | 1, 2 | 47 | |||
CR456482.1 | - | 14, 15 | 1851 | |||
KU178799.1 | - | 14, 15 | 1830 | |||
KU178800.1 | - | 15, 16 | 1991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
