siRNA Details
Gene Symbol: L3MBTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031488.4 | NP_113676.2 | 15 | 1909 | ||
XM_011530420.2 | XP_011528722.1 | 14 | 1892 | |||
XM_011530421.2 | XP_011528723.1 | 15 | 1978 | |||
XM_017028976.1 | XP_016884465.1 | 13 | 1686 | |||
XM_017028977.1 | XP_016884466.1 | 14 | 2205 | |||
XM_017028978.1 | XP_016884467.1 | 15 | 2160 | |||
XM_017028979.1 | XP_016884468.1 | 15 | 2105 | |||
XM_017028980.1 | XP_016884469.1 | 14 | 1792 | |||
GenBank mRNA | AJ305226.1 | - | 15 | 1894 | ||
AJ305227.1 | - | 16 | 1999 | |||
AK056443.1 | - | 15 | 1906 | |||
AK074091.1 | - | 13 | 4593 | |||
AL136564.1 | - | 14 | 2389 | |||
BC017191.2 | - | 15 | 1898 | 3138444 | ||
BU184979.1 | - | 2 | 72 | |||
CR456482.1 | - | 15 | 1876 | |||
KU178799.1 | - | 15 | 1855 | |||
KU178800.1 | - | 16 | 2016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
