siRNA Details
Gene Symbol: HNRNPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077442.1 | NP_001070910.1 | 8 | 1471 | ||
NM_001077443.1 | NP_001070911.1 | 8 | 1432 | |||
NM_004500.3 | NP_004491.2 | 9 | 1458 | |||
NM_031314.2 | NP_112604.2 | 9 | 1497 | |||
XM_006720125.2 | XP_006720188.1 | 9 | 2114 | |||
XM_011536708.1 | XP_011535010.1 | 10 | 1531 | |||
XM_011536709.2 | XP_011535011.1 | 9 | 1952 | |||
XM_011536710.2 | XP_011535012.1 | 9 | 2153 | |||
XM_011536711.2 | XP_011535013.1 | 9 | 2160 | |||
XM_011536712.1 | XP_011535014.1 | 10 | 1492 | |||
XM_017021252.1 | XP_016876741.1 | 9 | 2121 | |||
XM_017021253.1 | XP_016876742.1 | 9 | 1912 | |||
GenBank mRNA | AK126950.1 | - | 8 | 4166 | ||
AK223517.1 | - | 9 | 1436 | |||
AK302213.1 | - | 7 | 1494 | |||
AL833432.1 | - | 6 | 1035 | |||
BC003394.1 | - | 9 | 1389 | 3447724 | ||
BC066932.1 | - | 8 | 1377 | 4375450 | ||
BC092433.1 | - | 5 | 833 | |||
BC103758.1 | - | 8 | 1377 | 6187512 | ||
BX247961.1 | - | 8 | 1188 | |||
BX247992.1 | - | 8 | 1343 | |||
M16342.1 | - | 9 | 1334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
