siRNA Details
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005776.2 | NP_005767.1 | 4 | 409 | ||
GenBank mRNA | AF070654.1 | - | 4 | 362 | ||
AF104398.1 | - | 4 | 362 | |||
AK074781.1 | - | 4 | 314 | |||
AK311897.1 | - | 4 | 322 | |||
AY358635.1 | - | 4 | 319 | |||
BC103741.1 | - | 4 | 410 | 6025795 | ||
BG503618.1 | - | 3 | 211 | |||
BQ233425.1 | - | 409 | ||||
BQ937490.1 | - | 4 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
