siRNA Details
Gene Symbol: EIF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001969.4 | NP_001960.2 | 12 | 2703 | ||
NM_183004.4 | NP_892116.2 | 11 | 2560 | |||
GenBank mRNA | AK026933.1 | - | 12 | 2495 | ||
BC032866.2 | - | 11 | 2511 | 5261866 | ||
BX537367.1 | - | 10 | 2939 | |||
CR627002.1 | - | 1 | 1071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
