siRNA Details
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244249.1 | NP_001231178.1 | 5 | 1428 | ||
NM_001641.3 | NP_001632.2 | 5 | 1433 | |||
NM_080648.2 | NP_542379.1 | 5 | 1367 | |||
NM_080649.2 | NP_542380.1 | 5 | 1377 | |||
GenBank mRNA | AK098588.1 | - | 5 | 1317 | ||
AK291100.1 | - | 5 | 1342 | |||
BC002338.2 | - | 5 | 1318 | 2819505 | ||
BC004979.1 | - | 5 | 1230 | 2905681 | ||
BC008145.1 | - | 5 | 1316 | 4180383 | ||
BC019291.1 | - | 5 | 1318 | 2823545 | ||
BC095428.1 | - | 6 | 1307 | 30709297 | ||
BX161454.1 | - | 5 | 1267 | |||
D90373.1 | - | 5 | 1306 | |||
M80261.1 | - | 5 | 1223 | |||
M81955.1 | - | 5 | 1311 | |||
S43127.1 | - | 5 | 1324 | |||
U79268.1 | - | 5 | 1279 | |||
X59764.1 | - | 5 | 1310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
