siRNA Details
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244249.1 | NP_001231178.1 | 5 | 1138 | ||
NM_001641.3 | NP_001632.2 | 5 | 1143 | |||
NM_080648.2 | NP_542379.1 | 5 | 1077 | |||
NM_080649.2 | NP_542380.1 | 5 | 1087 | |||
GenBank mRNA | AK098588.1 | - | 5 | 1027 | ||
AK291100.1 | - | 5 | 1052 | |||
BC002338.2 | - | 5 | 1028 | 2819505 | ||
BC004979.1 | - | 5 | 940 | 2905681 | ||
BC008145.1 | - | 5 | 1026 | 4180383 | ||
BC019291.1 | - | 5 | 1028 | 2823545 | ||
BC095428.1 | - | 6 | 1017 | 30709297 | ||
BT007236.1 | - | 4 | 811 | |||
BT020133.1 | - | 4 | 811 | |||
BX161454.1 | - | 5 | 977 | |||
D90373.1 | - | 5 | 1016 | |||
M80261.1 | - | 5 | 933 | |||
M81955.1 | - | 5 | 1021 | |||
S43127.1 | - | 5 | 1034 | |||
U79268.1 | - | 5 | 989 | |||
X59764.1 | - | 5 | 1020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
