siRNA Details
Gene Symbol: SPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012444.2 | NP_036576.1 | 5 | 580 | ||
NM_198265.1 | NP_937998.1 | 4 | 466 | |||
XM_005260379.3 | XP_005260436.1 | 5 | 598 | |||
XM_005260380.3 | XP_005260437.1 | 5 | 598 | |||
XM_005260381.3 | XP_005260438.1 | 5 | 598 | |||
XM_005260382.4 | XP_005260439.1 | 5 | 598 | |||
XM_011528756.2 | XP_011527058.1 | 4 | 484 | |||
XM_011528757.2 | XP_011527059.1 | 5 | 432 | |||
GenBank mRNA | AF149310.1 | - | 5 | 520 | ||
AF169385.1 | - | 5 | 595 | |||
BC033591.1 | - | 4 | 428 | 5167214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
