siRNA Details
Gene Symbol: CRNKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278625.1 | NP_001265554.1 | 2 | 330 | ||
NM_001278626.1 | NP_001265555.1 | 2 | 330 | |||
NM_001278627.1 | NP_001265556.1 | 2 | 236 | |||
NM_016652.5 | NP_057736.4 | 2 | 366 | |||
GenBank mRNA | AF255443.2 | - | 2 | 330 | ||
AF318303.1 | - | 2 | 366 | |||
AF318304.1 | - | 2 | 330 | |||
AF318305.1 | - | 2 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
