siRNA Details
Gene Symbol: MROH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152503.5 | NP_689716.4 | 10 | 1360 | ||
NM_213631.2 | NP_998796.1 | 10 | 1360 | |||
NM_213632.2 | NP_998797.2 | 9 | 1255 | |||
GenBank mRNA | AK093432.1 | - | 9 | 1255 | ||
AL833864.1 | - | 3 | 257 | |||
BC030006.1 | - | 10 | 1337 | 4822338 | ||
BC057767.1 | - | 10 | 1343 | 5272440 | ||
BX098572.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
