siRNA Details
Gene Symbol: MROH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152503.5 | NP_689716.4 | 12 | 1729 | ||
NM_213631.2 | NP_998796.1 | 12 | 1729 | |||
NM_213632.2 | NP_998797.2 | 11 | 1624 | |||
GenBank mRNA | AK093432.1 | - | 11 | 1624 | ||
AL833864.1 | - | 4 | 473 | |||
BC030006.1 | - | 12 | 1706 | 4822338 | ||
BC057767.1 | - | 12 | 1712 | 5272440 | ||
BP371691.1 | - | 1 | 34 | |||
BX098572.1 | - | 4 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
