siRNA Details
Gene Symbol: EPPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020398.3 | NP_065131.1 | 4 | 1126 | ||
GenBank mRNA | AF286370.2 | - | 4 | 1078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
