siRNA Details
Gene Symbol: EPPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302861.1 | NP_001289790.1 | 4 | 730 | ||
NM_020398.3 | NP_065131.1 | 4 | 719 | |||
GenBank mRNA | AF286368.1 | - | 4 | 671 | ||
AF286369.1 | - | 3 | 1166 | |||
AF286370.2 | - | 4 | 671 | |||
AK126022.1 | - | 2 | 3967 | |||
AL118493.1 | - | 4 | 673 | |||
BC044829.1 | - | 4 | 682 | 5171493 | ||
BC053369.1 | - | 4 | 677 | 5165509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
