siRNA Details
Gene Symbol: NDFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161407.1 | NP_001154879.1 | 3 | 583 | ||
NM_019080.2 | NP_061953.2 | 3 | 583 | |||
XM_006719840.3 | XP_006719903.1 | 3 | 982 | |||
GenBank mRNA | AB032991.1 | - | 3 | 347 | ||
AB097019.1 | - | 3 | 555 | |||
AB097028.1 | - | 3 | 555 | |||
AB097029.1 | - | 3 | 555 | |||
AB097030.1 | - | 3 | 233 | |||
AB097031.1 | - | 3 | 233 | |||
AK001723.1 | - | 3 | 374 | |||
AK098708.1 | - | 4 | 336 | |||
AK294839.1 | - | 3 | 254 | |||
AK297197.1 | - | 3 | 290 | |||
BC021988.1 | - | 3 | 315 | 4101140 | ||
BC026126.2 | - | 3 | 344 | 3027907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
