siRNA Details
Gene Symbol: FGF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002010.2 | NP_002001.1 | 3 | 1942 | ||
XM_011534996.2 | XP_011533298.1 | 3 | 1271 | |||
GenBank mRNA | D14838.1 | - | 3 | 1284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
