siRNA Details
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005335.1 | NP_001005335.1 | 12, 13 | 1483 | ||
NM_001533.2 | NP_001524.2 | 12, 13 | 1717 | |||
XM_011526886.1 | XP_011525188.1 | 12, 13 | 1506 | |||
XM_011526887.1 | XP_011525189.1 | 12, 13 | 1550 | |||
XM_011526888.2 | XP_011525190.1 | 10, 11 | 1605 | |||
XM_011526889.1 | XP_011525191.1 | 12, 13 | 1601 | |||
GenBank mRNA | AB044547.1 | - | 12, 13 | 1712 | ||
AK092828.1 | - | 6, 7 | 2016 | |||
AK094966.1 | - | 5, 6 | 2707 | |||
AK292115.1 | - | 12, 13 | 1482 | |||
AK313652.1 | - | 12, 13 | 1652 | |||
BC069184.1 | - | 12, 13 | 1656 | |||
X16135.1 | - | 12, 13 | 1641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
