siRNA Details
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005335.1 | NP_001005335.1 | 13 | 1752 | ||
NM_001533.2 | NP_001524.2 | 13 | 1986 | |||
XM_011526886.1 | XP_011525188.1 | 13 | 1775 | |||
XM_011526887.1 | XP_011525189.1 | 13 | 1819 | |||
XM_011526888.2 | XP_011525190.1 | 11 | 1874 | |||
XM_011526889.1 | XP_011525191.1 | 13 | 1870 | |||
GenBank mRNA | AB044547.1 | - | 13 | 1978 | ||
AK092828.1 | - | 7 | 2282 | |||
AK094966.1 | - | 6 | 2974 | |||
AK097975.1 | - | 4 | 1991 | |||
AV743120.2 | - | 1 | 260 | |||
BC069184.1 | - | 13 | 1916 | |||
X16135.1 | - | 13 | 1905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
