siRNA Details
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001005335.1 | NP_001005335.1 | 8 | 939 | ||
NM_001533.2 | NP_001524.2 | 8 | 1173 | |||
XM_011526886.1 | XP_011525188.1 | 8 | 962 | |||
XM_011526887.1 | XP_011525189.1 | 8 | 1006 | |||
XM_011526888.2 | XP_011525190.1 | 6 | 1061 | |||
XM_011526889.1 | XP_011525191.1 | 8 | 1057 | |||
GenBank mRNA | AB044547.1 | - | 8 | 1168 | ||
AK092828.1 | - | 2 | 1472 | |||
AK094966.1 | - | 1 | 2163 | |||
AK292115.1 | - | 8 | 938 | |||
AK301062.1 | - | 10 | 921 | |||
AK313652.1 | - | 8 | 1108 | |||
AY927544.1 | - | LIT3550 AY927544 |
1 | 12 | ||
BC069184.1 | - | 8 | 1112 | |||
X16135.1 | - | 8 | 1097 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177301, CV561640, BC099683, BC027206, AK144684.
Gene Ontology Categories:
