siRNA Details
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146326.1 | NP_001139798.1 | 7, 8 | 743 | ||
NM_001146327.1 | NP_001139799.1 | 7, 8 | 830 | |||
NM_015363.4 | NP_056178.3 | 6, 7 | 667 | |||
GenBank mRNA | AF166122.1 | - | 6, 7 | 650 | ||
BC104876.1 | - | 6, 7 | 574 | 8143879 | ||
BC104878.1 | - | 6, 7 | 658 | 8143881 |
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 6, 7 | 980 | ||
NM_001146185.1 | NP_001139657.1 | 6, 7 | 666 | |||
NM_001146186.1 | NP_001139658.1 | 4, 5 | 1013 | |||
NM_001146187.1 | NP_001139659.1 | 6, 7 | 669 | |||
NM_006210.2 | NP_006201.1 | 7, 8 | 1056 | |||
GenBank mRNA | AB003039.1 | - | 6, 7 | 630 | ||
AB006625.2 | - | 6, 7 | 642 | |||
AF208967.1 | - | 7, 8 | 1026 | |||
AF208968.1 | - | 6, 7 | 950 | |||
AF208969.1 | - | 6, 7 | 636 | |||
AF208970.1 | - | 4, 5 | 1014 | |||
AK295679.1 | - | 6, 7 | 669 | |||
AK307476.1 | - | 6, 7 | 669 | |||
BC052616.1 | - | 4, 5 | 658 | |||
BC136268.1 | - | 5, 6 | 497 | 9020255 | ||
BC150272.1 | - | 6, 7 | 642 | 9007244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
