siRNA Details
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146326.1 | NP_001139798.1 | 8 | 797 | ||
NM_001146327.1 | NP_001139799.1 | 8 | 884 | |||
NM_015363.4 | NP_056178.3 | 7 | 721 | |||
GenBank mRNA | AF166122.1 | - | 7 | 704 | ||
BC104876.1 | - | 7 | 628 | 8143879 | ||
BC104878.1 | - | 7 | 712 | 8143881 |
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 7 | 1034 | ||
NM_001146185.1 | NP_001139657.1 | 7 | 720 | |||
NM_001146186.1 | NP_001139658.1 | 5 | 1067 | |||
NM_001146187.1 | NP_001139659.1 | 7 | 723 | |||
NM_006210.2 | NP_006201.1 | 8 | 1110 | |||
GenBank mRNA | AB006625.2 | - | 7 | 696 | ||
AF208967.1 | - | 8 | 1080 | |||
AF208968.1 | - | 7 | 1004 | |||
AF208969.1 | - | 7 | 690 | |||
AF208970.1 | - | 5 | 1068 | |||
AK295679.1 | - | 7 | 723 | |||
AK307476.1 | - | 7 | 723 | |||
BC052616.1 | - | 5 | 712 | |||
BC136268.1 | - | 6 | 551 | 9020255 | ||
BC150272.1 | - | 7 | 696 | 9007244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
