siRNA Details
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146326.1 | NP_001139798.1 | 4, 5 | 477 | ||
NM_001146327.1 | NP_001139799.1 | 4, 5 | 564 | |||
NM_015363.4 | NP_056178.3 | 3, 4 | 401 | |||
GenBank mRNA | AF166122.1 | - | 3, 4 | 384 | ||
BC104876.1 | - | 3, 4 | 308 | 8143879 | ||
BC104878.1 | - | 3, 4 | 392 | 8143881 | ||
DC400394.1 | - | 3, 4 | 460 |
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 3, 4 | 717 | ||
NM_001146185.1 | NP_001139657.1 | 3, 4 | 403 | |||
NM_001146186.1 | NP_001139658.1 | 1, 2 | 750 | |||
NM_001146187.1 | NP_001139659.1 | 3, 4 | 403 | |||
NM_006210.2 | NP_006201.1 | 4, 5 | 793 | |||
GenBank mRNA | AB003039.1 | - | 3, 4 | 364 | ||
AB006625.2 | - | 3, 4 | 376 | |||
AF208967.1 | - | 4, 5 | 763 | |||
AF208968.1 | - | 3, 4 | 687 | |||
AF208969.1 | - | 3, 4 | 373 | |||
AF208970.1 | - | 1, 2 | 751 | |||
AK295679.1 | - | 3, 4 | 403 | |||
AK307476.1 | - | 3, 4 | 403 | |||
BC052616.1 | - | 1, 2 | 392 | |||
BC136268.1 | - | 2, 3 | 231 | 9020255 | ||
BC150272.1 | - | 3, 4 | 376 | 9007244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
