siRNA Details
Gene Symbol: CAPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004058.4 | NP_004049.2 | 4 | 1134 | ||
GenBank mRNA | AK090469.1 | - | 4 | 3418 | ||
AK226026.1 | - | 850 | ||||
BC011961.1 | - | 2 | 373 | |||
BC080599.1 | - | 4 | 542 | 4876766 | ||
BC101513.1 | - | 3 | 471 | 8069019 | ||
BC104766.1 | - | 3 | 471 | 8143769 | ||
BG768134.1 | - | 4 | 572 | |||
X97966.1 | - | 3 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
