siRNA Details
Gene Symbol: CAPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004058.4 | NP_004049.2 | 5 | 1718 | ||
NM_080590.3 | NP_542157.2 | 5 | 1637 | |||
GenBank mRNA | AK090469.1 | - | 5 | 4002 | ||
AK226026.1 | - | 1434 | ||||
BC011961.1 | - | 3 | 957 | |||
BC019065.1 | - | 1294 | ||||
BC080599.1 | - | 5 | 1126 | 4876766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
