siRNA Details
Gene Symbol: GLCCI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138426.3 | NP_612435.1 | 5, 6 | 1581 | ||
GenBank mRNA | AL833694.1 | - | 1, 2 | 76 | ||
BC050291.1 | - | 5, 6 | 1532 | 5272003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
