siRNA Details
Gene Symbol: GLCCI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138426.3 | NP_612435.1 | 8 | 2951 | ||
GenBank mRNA | AK093655.1 | - | 1 | 284 | ||
AK126731.1 | - | 2 | 2100 | |||
AL832860.1 | - | 2 | 1063 | |||
AL833694.1 | - | 4 | 1445 | |||
BC050291.1 | - | 8 | 2900 | 5272003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
