siRNA Details
Gene Symbol: ZNF510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314059.1 | NP_001300988.1 | 6 | 3331 | ||
NM_001314060.1 | NP_001300989.1 | 5 | 2883 | |||
NM_014930.2 | NP_055745.1 | 6 | 2942 | |||
XM_005251808.2 | XP_005251865.1 | 6 | 2929 | |||
XM_011518393.2 | XP_011516695.1 | 6 | 3387 | |||
XM_017014483.1 | XP_016869972.1 | 7 | 2994 | |||
GenBank mRNA | AB023189.1 | - | 5 | 2859 | ||
AK292600.1 | - | 6 | 2912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
