siRNA Details
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002269.2 | NP_001002269.1 | 2 | 485 | ||
NM_016042.3 | NP_057126.2 | 2 | 485 | |||
GenBank mRNA | AF151860.1 | - | 2 | 411 | ||
AF229833.1 | - | 2 | 182 | |||
AF281132.1 | - | 2 | 461 | |||
AK289571.1 | - | 2 | 454 | |||
AK290864.1 | - | 2 | 450 | |||
BC002437.2 | - | 2 | 449 | 3346075 | ||
BC008880.2 | - | 2 | 439 | 4300826 | ||
BP237222.1 | - | 2 | 484 | |||
CF994051.1 | - | 2 | 493 | |||
DA352003.1 | - | 2 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
