siRNA Details
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016042.3 | NP_057126.2 | 3 | 658 | ||
GenBank mRNA | AF151860.1 | - | 3 | 584 | ||
AF281132.1 | - | 3 | 634 | |||
AK290864.1 | - | 3 | 623 | |||
BC002437.2 | - | 3 | 622 | 3346075 | ||
BC008880.2 | - | 3 | 612 | 4300826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
