siRNA Details
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024326.3 | NP_077302.3 | 4 | 2239 | ||
XM_005270149.2 | XP_005270206.1 | 6 | 1583 | |||
XM_005270150.4 | XP_005270207.1 | 5 | 2033 | |||
XM_005270151.3 | XP_005270208.1 | 5 | 1189 | |||
XM_017016631.1 | XP_016872120.1 | 5 | 1320 | |||
GenBank mRNA | BC002912.2 | - | 4 | 1056 | 3944940 | |
BC036120.1 | - | 4 | 2220 | 5286464 | ||
BC130566.1 | - | 5 | 1292 | 40146597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
