siRNA Details
Gene Symbol: ANKRD26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256053.1 | NP_001242982.1 | 32 | 5071 | ||
NM_014915.2 | NP_055730.2 | 32 | 5074 | |||
XM_006717423.2 | XP_006717486.1 | 33 | 6160 | |||
XM_006717425.3 | XP_006717488.1 | 33 | 6160 | |||
XM_011519416.2 | XP_011517718.1 | 33 | 6160 | |||
XM_017015928.1 | XP_016871417.1 | 33 | 6160 | |||
XM_017015929.1 | XP_016871418.1 | 34 | 6148 | |||
XM_017015930.1 | XP_016871419.1 | 33 | 6160 | |||
XM_017015931.1 | XP_016871420.1 | 33 | 6160 | |||
XM_017015932.1 | XP_016871421.1 | 33 | 6160 | |||
XM_017015933.1 | XP_016871422.1 | 34 | 6368 | |||
GenBank mRNA | AB028997.1 | - | 32 | 5049 | ||
AL137351.1 | - | 5 | 892 | |||
AL162063.1 | - | 1 | 50 | |||
BC110646.1 | - | 2 | 267 | |||
BC130453.1 | - | 32 | 5015 | 40146484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
