siRNA Details
Gene Symbol: HNF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000458.3 | NP_000449.1 | 2, 3 | 734 | ||
XM_011525160.1 | XP_011523462.1 | 2, 3 | 715 | |||
XM_011525161.1 | XP_011523463.1 | 2, 3 | 715 | |||
XM_011525162.2 | XP_011523464.1 | 2, 3 | 715 | |||
XM_011525163.2 | XP_011523465.1 | 2, 3 | 715 | |||
GenBank mRNA | AK290131.1 | - | 2, 3 | 715 | ||
AK298393.1 | - | 3, 4 | 584 | |||
BC017714.1 | - | 2, 3 | 728 | 4414015 | ||
BT007126.1 | - | 2, 3 | 540 | |||
CR536572.1 | - | 2, 3 | 540 | |||
HM116555.1 | - | 2, 3 | 674 | |||
X58840.1 | - | 2, 3 | 734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
