siRNA Details
Gene Symbol: HNF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000458.3 | NP_000449.1 | 5 | 1329 | ||
NM_001165923.3 | NP_001159395.1 | 5 | 1251 | |||
NM_001304286.1 | NP_001291215.1 | 5 | 1251 | |||
XM_011525160.1 | XP_011523462.1 | 5 | 1310 | |||
XM_011525161.1 | XP_011523463.1 | 5 | 1310 | |||
XM_011525164.1 | XP_011523466.1 | 5 | 1232 | |||
GenBank mRNA | AK290131.1 | - | 5 | 1310 | ||
AK296633.1 | - | 5 | 1232 | |||
AK298393.1 | - | 6 | 1179 | |||
BC017714.1 | - | 5 | 1323 | 4414015 | ||
BT007126.1 | - | 5 | 1135 | |||
CR536572.1 | - | 5 | 1135 | |||
HM116553.1 | - | 5 | 1191 | |||
HM116554.1 | - | 5 | 1172 | |||
HM116555.1 | - | 5 | 1269 | |||
HM116556.1 | - | 5 | 1191 | |||
X58840.1 | - | 5 | 1329 |

More Information
Other Species Targeted

Gene Ontology Categories:
