siRNA Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035512.1 | NP_001030589.1 | 2, 3 | 109 | ||
NM_001278172.1 | NP_001265101.1 | 2, 3 | 109 | |||
GenBank mRNA | AB211234.1 | - | 2, 3 | 103 | ||
AB212048.1 | - | 2, 3 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
