siRNA Details
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287437.1 | NP_001274366.1 | 11 | 3540 | ||
NM_001287439.1 | NP_001274368.1 | 11 | 3694 | |||
NM_001287440.1 | NP_001274369.1 | 11 | 3491 | |||
NM_001287442.1 | NP_001274371.1 | 11 | 3448 | |||
NM_001287443.1 | NP_001274372.1 | 11 | 3452 | |||
NM_199320.3 | NP_955352.1 | 11 | 3576 | |||
XM_005263232.1 | XP_005263289.1 | 11 | 3418 | |||
XM_017008626.1 | XP_016864115.1 | 10 | 3415 | |||
XM_017008627.1 | XP_016864116.1 | 10 | 3462 | |||
GenBank mRNA | AB058710.1 | - | 7 | 2876 | ||
AK026132.1 | - | 2 | 1692 | |||
AK127326.1 | - | 11 | 3348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
