siRNA Details
Gene Symbol: GAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018983.3 | NP_061856.1 | 3 | 581 | ||
NM_032993.2 | NP_127460.1 | 3 | 322 | |||
GenBank mRNA | AJ276003.1 | - | 3 | 538 | ||
AK310489.1 | - | 3 | 464 | |||
AY780787.1 | - | 2 | 274 | |||
BC003413.1 | - | 3 | 322 | 3451136 | ||
CN428426.1 | - | 3 | 546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
