siRNA Details
Gene Symbol: GAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018983.3 | NP_061856.1 | 5 | 780 | ||
NM_032993.2 | NP_127460.1 | 5 | 521 | |||
GenBank mRNA | AJ276003.1 | - | 5 | 737 | ||
AY780787.1 | - | 4 | 473 | |||
BC003413.1 | - | 5 | 521 | 3451136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
