siRNA Details
Gene Symbol: C11orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024806.3 | NP_079082.2 | 2 | 848 | ||
NM_199124.2 | NP_954575.1 | 2 | 848 | |||
XM_005271680.4 | XP_005271737.1 | 2 | 709 | |||
XM_006718915.2 | XP_006718978.1 | 2 | 583 | |||
XM_011543009.2 | XP_011541311.1 | 2 | 917 | |||
XM_017018338.1 | XP_016873827.1 | 2 | 583 | |||
XM_017018339.1 | XP_016873828.1 | 2 | 616 | |||
XM_017018340.1 | XP_016873829.1 | 2 | 427 | |||
XM_017018341.1 | XP_016873830.1 | 2 | 1198 | |||
XM_017018342.1 | XP_016873831.1 | 2 | 583 | |||
XM_017018343.1 | XP_016873832.1 | 2 | 583 | |||
GenBank mRNA | AK027207.1 | - | 1 | 202 | ||
AK097534.1 | - | 2 | 693 | |||
AK291625.1 | - | 2 | 451 | |||
BC009820.2 | - | 1 | 254 | |||
BC068507.1 | - | 2 | 611 | 5298472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
