siRNA Details
Gene Symbol: ZNF268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165881.2 | NP_001159353.1 | 6 | 699 | ||
NM_001165882.2 | NP_001159354.1 | 6 | 706 | |||
NM_001165883.1 | NP_001159355.1 | 4 | 600 | |||
NM_001165884.2 | NP_001159356.2 | 3 | 399 | |||
NM_001165885.1 | NP_001159357.1 | 5 | 696 | |||
NM_001165886.1 | NP_001159358.1 | 4 | 495 | |||
NM_001165887.1 | NP_001159359.1 | 6 | 728 | |||
NM_003415.2 | NP_003406.1 | 6 | 823 | |||
NM_152943.2 | NP_694422.2 | 7 | 929 | |||
GenBank mRNA | AF317549.1 | - | 6 | 823 | ||
AF385187.1 | - | 7 | 779 | |||
AF432217.1 | - | 3 | 299 | |||
AK027712.1 | - | 7 | 843 | |||
AY583691.1 | - | 7 | 715 | |||
BC110542.2 | - | 6 | 699 | 40036577 | ||
CN255799.1 | - | 6 | 683 | |||
DB079580.1 | - | 3 | 426 | |||
DQ057356.1 | - | 5 | 485 | |||
DQ057357.1 | - | 4 | 284 | |||
DQ057358.1 | - | 4 | 389 | |||
DQ057359.1 | - | 5 | 390 | |||
DQ057360.1 | - | 6 | 517 | |||
DQ057361.1 | - | 3 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
