siRNA Details
Gene Symbol: H2AFJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177925.3 | NP_808760.1 | 1 | 160 | ||
NR_027716.2 | 1 | 160 | ||||
GenBank mRNA | AK001765.1 | - | 1 | 91 | ||
AK303301.1 | - | 1, 2 | 91 | |||
BC003602.1 | - | 1 | 81 | 3354610 | ||
BM689354.1 | - | 1 | 164 | |||
BU594879.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
