siRNA Details
Gene Symbol: H2AFJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177925.3 | NP_808760.1 | 1 | 183 | ||
NR_027716.2 | 1 | 183 | ||||
GenBank mRNA | AK001765.1 | - | 1 | 114 | ||
BC003602.1 | - | 1 | 104 | 3354610 | ||
BM689354.1 | - | 1 | 187 | |||
BU594879.1 | - | 1 | 161 | |||
DA808847.1 | - | 1 | 184 | |||
DB084616.1 | - | 1 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
