siRNA Details
Gene Symbol: H2AFJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177925.3 | NP_808760.1 | 1 | 241 | ||
NR_027716.2 | 1 | 241 | ||||
GenBank mRNA | AK001765.1 | - | 1 | 172 | ||
AK303301.1 | - | 2 | 118 | |||
BC003602.1 | - | 1 | 162 | 3354610 | ||
BM689354.1 | - | 1 | 245 | |||
BU594879.1 | - | 1 | 219 | |||
DA808847.1 | - | 1 | 242 | |||
DB084616.1 | - | 1 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
