siRNA Details
Gene Symbol: NANOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297698.1 | NP_001284627.1 | 4 | 1718 | ||
NM_024865.3 | NP_079141.2 | 4 | 1766 | |||
GenBank mRNA | AB093576.1 | - | 4 | 1779 | ||
AK022643.1 | - | 4 | 1779 | |||
BC069807.1 | - | 1 | 1716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
