siRNA Details
Gene Symbol: LY6G5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025262.3 | NP_079538.3 | 3 | 394 | ||
GenBank mRNA | AA179679.1 | - | 1 | 36 | ||
AJ315540.1 | - | 4 | 498 | |||
AJ315541.1 | - | 4 | 618 | |||
AJ315542.1 | - | 3 | 385 | |||
AJ315543.1 | - | 3 | 479 | |||
AJ315544.1 | - | 2 | 1369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
