siRNA Details
Gene Symbol: LY6G5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025262.3 | NP_079538.3 | 2 | 134 | ||
GenBank mRNA | AJ315540.1 | - | 3 | 238 | ||
AJ315541.1 | - | 3 | 358 | |||
AJ315542.1 | - | 2 | 125 | |||
AJ315543.1 | - | 2 | 219 | |||
AJ315544.1 | - | 1 | 1109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
