siRNA Details
Gene Symbol: RNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003958.3 | NP_003949.1 | 8 | 4496 | ||
NM_183078.2 | NP_898901.1 | 7 | 4291 | |||
NR_046399.1 | 8 | 4602 | ||||
GenBank mRNA | AB014546.1 | - | 8 | 4415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
