siRNA Details
Gene Symbol: ZBTB24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014797.2 | NP_055612.2 | 4 | 1303 | ||
GenBank mRNA | AB007901.1 | - | 4 | 1303 | ||
AK309697.1 | - | 4 | 1257 | |||
BC117374.1 | - | 3 | 1171 | 40125925 | ||
BC117376.1 | - | 3 | 1171 | 40125927 | ||
BX648883.1 | - | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
