siRNA Details
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003190.4 | NP_003181.3 | 8 | 3144 | ||
NM_172209.2 | NP_757346.2 | 7 | 2883 | |||
XM_011514828.2 | XP_011513130.1 | 6 | 2886 | |||
XM_017011227.1 | XP_016866716.1 | 7 | 2976 | |||
GenBank mRNA | AF029750.1 | - | 8 | 2839 | ||
AY007154.1 | - | 1 | 573 | |||
BC064966.1 | - | 7 | 2271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
